Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIF18A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558728
Description
Immunogen sequence: LHCHHLHLQN KDLKAQIRHM MDLACLQEQQ HRQTEAVLNA LLPTLRKQYC TLKEAGLSNA AFESDFKEIE HLVERKKVVV WADQTAEQPK QND Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 82%.
May have a role in a signaling pathway. Could act as a tumor suppressor.
Specifications
KIF18A | |
Polyclonal | |
Unconjugated | |
KIF18A | |
AU024633; B130001M12Rik; Kif18a; kinesin family member 18A; kinesin superfamily protein 18A; kinesin-like protein KIF18A; Marrow stromal KIF18A; MS-KIF18A; N-8 kinesin; OK/SW-cl.108; PPP1R99; protein phosphatase 1, regulatory subunit 99 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
81930 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NI77 | |
KIF18A | |
Recombinant protein corresponding to Human KIF18A. Recombinant protein control fragment (Product #RP-97337). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction