Learn More
KIF18A Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301079A
Description
The recommended shelf life for this product is 1 year from date of receipt.
KIF18A is a member of the kinesin superfamily of microtubule-associated molecular motors (see MIM 148760) that use hydrolysis of ATP to produce force and movement along microtubules (Luboshits and Benayahu, 2005 [PubMed 15878648]).Specifications
| KIF18A | |
| Polyclonal | |
| Unconjugated | |
| KIF18A | |
| MS-KIF18A, kinesin-like protein KIF18A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 81930 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q8NI77 | |
| KIF18A | |
| Between 650 and 700 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.