Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIF18B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555014
Description
Immunogen sequence: STLQVVVRVR PPTPRELDSQ RRPVVQVVDE RVLVFNPEEP DGGFPGLKWG GTHDGPKKKG KDLTFVFDRV FGEAATQQDV FQHTTHSVLD SFLQGYN Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 82%.
In complex with KIF2C, constitutes the major microtubule plus-end depolymerizing activity in mitotic cells. Its major role may be to transport KIF2C and/or MAPRE1 along microtubules. [UniProt].
Specifications
KIF18B | |
Polyclonal | |
Unconjugated | |
KIF18B | |
KIF18B; kinesin family member 18B; kinesin-like protein KIF18B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
146909 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86Y91 | |
KIF18B | |
Recombinant protein corresponding to Human KIF18B. Recombinant protein control fragment (Product #RP-93540). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction