Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567284
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567284 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567284 Supplier Invitrogen™ Supplier No. PA567284
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SGDSKDQQLMV ALRVRPISVA ELEEGATLIA HKVDEQMVVL MDPMEDPDDI LRAHRSREKS YLFDVAFDFT ATQEMVYQ Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.

Plus end-directed microtubule-dependent motor protein that regulates the length of motile cilia by mediating depolymerization of microtubules at ciliary tips.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF19
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIF19
Gene Accession No. Q2TAC6
Gene Alias KIF19; Kif19a; KIF19A-like protein; kinesin family member 19; kinesin family member 19A; kinesin-like protein KIF19; N-8 kinesin; oocyte-G1; RGD1559936
Gene Symbols KIF19
Host Species Rabbit
Immunogen Recombinant Human KIF19. Recombinant protein control fragment (Product #RP-107922).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124602
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.