Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589493
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589493 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589493 Supplier Invitrogen™ Supplier No. PA589493
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKDSGDSKDQ QLMVALRVRP ISVAELEEGA TLIAHKVDEQ MVVLMDPMED PDDILRAHRS REKSYLFDVA FDFTATQEMV YQATTKSLIE GVISGYNATV FAYGPTGCGK TYTMLGTDQE PGIYVQTLND LFRAIEETSN DMEYEVSMSY LEIYNEMIRD LLNPSLGYLE LREDSKGVIQ VAGITEVSTI NAKEIMQLLM KGNRQRTQEP TAANQTSSRS HAVLQVTVRQ RSRVKNILQE VRQGRLFMID LAGSERASQT QNRGQRMKEG AHINRSLLAL GNCINALSDK GSNKYINYRD; Positive Samples: U-87MG, Mouse testis, Mouse lung, Rat lung; Cellular Location: Cell projection, Cytoplasm, cilium, cytoskeleton.

Plus end-directed microtubule-dependent motor protein that regulates the length of motile cilia by mediating depolymerization of microtubules at ciliary tips.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF19
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene KIF19
Gene Accession No. Q2TAC6
Gene Alias KIF19; Kif19a; KIF19A-like protein; kinesin family member 19; kinesin family member 19A; kinesin-like protein KIF19; N-8 kinesin; oocyte-G1; RGD1559936
Gene Symbols KIF19
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human KIF19 (NP_6949412).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124602, 303659
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.