Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540801
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540801 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540801 Supplier Invitrogen™ Supplier No. PA540801
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TTIVNPKQPK ETPKSFSFDY SYWSHTSPED INYASQKQVY RDIGEEMLQH Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 79%; Rat: 100%; Yeast: 100%; Zebrafish: 79%.

KIF1A is the motor for anterograde axonal transport of synaptic vesicle precursors.The protein encoded by this gene is a member of the kinesin family. This protein is highly similar to mouse heavy chain kinesin member 1A protein which is an anterograde motor protein that transports membranous organelles along axonal microtubules. It is thought that this protein may play a critical role in the development of axonal neuropathies resulting from impaired axonal transport. There are multiple polyadenylation sites found in this gene. Sequence Note: X90840.1 is a chimeric sequence. Only the ATSV region was propagated into this RefSeq record. [6/26/03, RefSeq staff]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF1A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KIF1A
Gene Accession No. Q12756
Gene Alias ATSV; axonal transporter of synaptic vesicles; C2orf20; C630002N23Rik; DKFZp686I2094; FLJ30229; Gm1626; HSN2C; hUnc-104; KIF1A; kinesin family member 1A; kinesin heavy chain member 1A; kinesin, heavy chain, member 1A, homolog of mouse; kinesin-like protein KIF1A; Kns1; MGC133285; MGC133286; Microtubule-based motor KIF1A; MRD9; N-3 kinesin; neuron-specific kinesin motor KIF1A; SPG30; tmp_locus_27; UNC104; unc-104- and KIF1A-related protein; Unknown (protein for IMAGE:7984457)
Gene Symbols KIF1A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human KIF1A (aa 35-84).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 547
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.