Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF1B Polyclonal Antibody

KIF1B Polyclonal Antibody

Supplier:  Thermo Scientific PA593117

Catalog No. PIPA593117


Only null left
Add to cart

Description

Description

KIF1B Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

KIF1B
Polyclonal
Unconjugated
KIF1B
A530096N05Rik; AI448212; AI506502; CMT2; CMT2A; CMT2A1; D4Mil1e; HMSNII; KIAA0591; KIAA1448; Kif1b; KIF1Bp130; KIF1Bp204; kinesin family member 1B; kinesin heavy chain member 1B; kinesin superfamily protein KIF1B; kinesin-family protein KIF1Bbeta3; kinesin-like protein KIF1B; Klp; N-3 kinesin; NBLST1
Rabbit
Affinity Chromatography
RUO
117548, 16561, 23095
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.38 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O60333, O88658, Q60575
KIF1B
Recombinant fusion protein containing a sequence corresponding to amino acids 1501-1770 of human KIF1B (NP_0558892).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.