Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIF1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | KIF1B |
---|---|
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
KIF1B Polyclonal specifically detects KIF1B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
KIF1B | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CMT2, CMT2A, CMT2A1, HMSNII, KIAA0591FLJ23699, KIAA1448, kinesin family member 1B, kinesin superfamily protein KIF1B, kinesin-like protein KIF1B, Klp, MGC134844, NBLST1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1501-1770 of human KIF1B (NP_055889.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Stem Cell Markers | |
PBS with 50% glycerol, pH7.3. | |
23095 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title