Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIF2 Mouse anti-Mouse, Unconjugated, Clone: 2B12.C1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT153625UG
Description
KIF2 Monoclonal antibody specifically detects KIF2 in Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blotting.
Specifications
KIF2 | |
Monoclonal | |
Unconjugated | |
Kinesin-2;Kinesin-like protein KIF2A | |
Mouse | |
79.75 kDa calculated. | |
25 μg | |
Primary | |
Mouse | |
Purified |
Immunohistochemistry (Paraffin), Western Blot | |
2B12.C1 | |
NP_001139251.1 | |
Kif2a, Kif2, Kns2 | |
T7-tagged full-length mouse recombinant Kinesin-like protein KIF2A. | |
Protein G Purified | |
Signaling | |
Clone 2B12.C1 is a mouse monoclonal antibody that detects Kinesin-like protein KIF2A in murine species. | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction