Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIF20A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583359
Description
Immunogen sequence: PLLSNEVIWL DSKQIRQEEM KKLSLLNGGL QEEELSTSLK RSVYIESRIG TSTSFDSGIA GLSSISQCTS SSQLDETSHR WAQPDTAPLP VPA.
Mitotic kinesin required for chromosome passenger complex (CPC)-mediated cytokinesis. Following phosphorylation by PLK1, involved in recruitment of PLK1 to the central spindle. Interacts with guanosine triphosphate (GTP)-bound forms of RAB6A and RAB6B. May act as a motor required for the retrograde RAB6 regulated transport of Golgi membranes and associated vesicles along microtubules. Has a microtubule plus end-directed motility. [UniProt].
Specifications
KIF20A | |
Polyclonal | |
Unconjugated | |
KIF20A | |
AA415432; GG10_2; KIF20A; kinesin family member 20A; kinesin-like protein 174; Kinesin-like protein KIF20A; mitotic kinesin-like protein 2; MKlp2; RAB6 interacting, kinesin-like (rabkinesin6); rab6-interacting kinesin-like protein; RAB6KIFL; Rabkinesin-6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10112 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95235 | |
KIF20A | |
Recombinant protein corresponding to Human KIF20A. Recombinant protein control fragment (Product #RP-96872). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction