Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIF21A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP158143
Description
KIF21A Polyclonal specifically detects KIF21A in Human samples. It is validated for Western Blot.Specifications
KIF21A | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
CFEOM1, CFEOM3B, FEOM1, FEOM3A, fibrosis of the extraocular muscles, congenital, 1, FLJ20052, KIAA1708DKFZp779C159, KIF2, kinesin family member 21A, Kinesin-like protein KIF2, kinesin-like protein KIF21A, Renal carcinoma antigen NY-REN-62 | |
Rabbit | |
186 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q7Z4S6 | |
KIF21A | |
Synthetic peptides corresponding to KIF21A (kinesin family member 21A) The peptide sequence was selected from the N terminal of KIF21A. Peptide sequence KEKRKKKSVAGKEDNTDTDQEKKEEKGVSERENNELEVEESQEVSDHEDE. | |
Affinity purified | |
RUO | |
55605 | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction