Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF21A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563572
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563572 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563572 Supplier Invitrogen™ Supplier No. PA563572
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QKEAQIKVLE GRLKQTEITS ATQNQLLFHM LKEKAELNPE LDALLGHALQ DLDSVPLENV EDSTDEDAPL NS Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

KIF21A gene ontology annotations related to this gene include microtubule-based movement.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF21A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Kif21a
Gene Accession No. Q7Z4S6
Gene Alias CFEOM1; FEOM1; FEOM3A; FLJ20052; KIAA1708; KIF2; Kif21a; kinesin family member 21A; kinesin-like protein KIF2; Kinesin-like protein KIF21A; N-5 kinesin; putative LOC540076 protein; renal carcinoma antigen NY-REN-62; Unknown (protein for IMAGE:8217042)
Gene Symbols Kif21a
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIF21A. Recombinant protein control fragment (Product #RP-103331).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55605
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.