Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF21A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735627 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5627 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5627 Supplier Proteintech Group Inc Supplier No. 272761AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KIF21A gene ontology annotations related to this gene include microtubule-based movement.

Specifications

Antigen KIF21A
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene KIF21A
Gene Accession No. Q7Z4S6, Q9QXL2
Gene Alias CFEOM, CFEOM1, FEOM, FEOM1, KIAA1708, KIF2, KIF21A, kinesin family member 21A, Kinesin like protein KIF2, Kinesin like protein KIF21A
Gene Symbols KIF21A
Host Species Rabbit
Immunogen KIF21A Fusion Protein Ag26192
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16564, 55605
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.