Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF21B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555556
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555556 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555556 Supplier Invitrogen™ Supplier No. PA555556
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RKTQEVSALR RLAKPMSERV AGRAGLKPPM LDSGAEVSAS TTSSEAESGA RSVSSIVRQW NRKINHFLGD HPAPTVNGTR PARKKFQKKG Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.

KIF6 belongs to the kinesin-like protein family. There are four named isoforms. The KIF6 719Arg variant has been associated with an increased risk of coronary disease and an increased response to statin therapy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF21B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIF21B
Gene Accession No. O75037
Gene Alias 2610511N21Rik; DKFZP434J212; KIAA0449; KIF21B; Kif6; kinesin family member 21B; kinesin-like protein KIF21B; Kinesin-like protein KIF6; LOW QUALITY PROTEIN: kinesin-like protein KIF21B; mKIAA0449; N-5 kinesin; zmp:0000000534
Gene Symbols KIF21B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIF21B. Recombinant protein control fragment (Product #RP-94091).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23046
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.