Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIF23 Recombinant Rabbit Monoclonal Antibody (9V4Y8)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535603
Description
Immunogen sequence: NAPPIRLRHR RSRSAGDRWV DHKPASNMQT ETVMQPHVPH AITVSVANEK ALAKCEKYML THQELASDGE IETKLIKGDI YKTRGGGQSV QFTDIETLKQ ESPNGSRKRR SSTVAPAQPD GAESEWTDVE TRCSVAVEMR AGSQLGPGYQ HHAQPKRKKP.
The protein encoded by this gene is a member of kinesin-like protein family. This family includes microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. This protein has been shown to cross-bridge antiparallel microtubules and drive microtubule movement in vitro. Alternate splicing of this gene results in two transcript variants encoding two different isoforms.
Specifications
KIF23 | |
Recombinant Monoclonal | |
1.58 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q02241 | |
KIF23 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 801-960 of human KIF23 (Q02241). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
ELISA, Immunoprecipitation, Western Blot | |
9V4Y8 | |
Unconjugated | |
KIF23 | |
3110001D19Rik; C87313; cb738; CHO1; KIF23; kif23 protein; kinesin family member 23; kinesin-like 5; kinesin-like 5 (mitotic kinesin-like protein 1); Kinesin-like protein 5; Kinesin-like protein KIF23; Kinesin-like protein KIF23-like protein; knsl5; Mitotic kinesin-like protein 1; mklp1; MKLP-1; wu:fi30e02; wu:fi39f08; zMKLP1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
315740, 9493 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction