Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100863
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100863 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100863 Supplier Invitrogen™ Supplier No. PA5100863
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KIF23.

The protein encoded by this gene is a member of kinesin-like protein family. This family includes microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. This protein has been shown to cross-bridge antiparallel microtubules and drive microtubule movement in vitro. Alternate splicing of this gene results in two transcript variants encoding two different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF23
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KIF23
Gene Accession No. E9Q5G3, Q02241
Gene Alias 3110001D19Rik; C87313; cb738; CHO1; KIF23; kif23 protein; kinesin family member 23; kinesin-like 5; kinesin-like 5 (mitotic kinesin-like protein 1); Kinesin-like protein 5; Kinesin-like protein KIF23; Kinesin-like protein KIF23-like protein; knsl5; Mitotic kinesin-like protein 1; mklp1; MKLP-1; wu:fi30e02; wu:fi39f08; zMKLP1
Gene Symbols KIF23
Host Species Rabbit
Immunogen A synthesized peptide derived from human KIF23(Accession Q02241), corresponding to amino acid residues V432-S482.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 71819, 9493
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.