Learn More
KIF24 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA554253
Description
Immunogen sequence: ASWLYECLCE AELAQYYSHF TALGLQKIDE LAKITMKDYS KLGVHDMNDR KRLFQLIKII KIMQEEDKAV SIPERHLQTS SLRIKSQELR SGPRRQLNFD SPADNKDRNA SNDGFEMCSL SDFSANEQKS T Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 82%.
Specifications
| KIF24 | |
| Polyclonal | |
| Unconjugated | |
| KIF24 | |
| C9orf48, bA571F15.4, kinesin-like protein KIF24 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 347240 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5T7B8 | |
| KIF24 | |
| Recombit protein corresponding to Human KIF24 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only