Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIF25 Antibody (OTI2F4), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271077H
Description
KIF25 Monoclonal specifically detects KIF25 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
KIF25 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
KIF25 | |
Full length recombinant protein of human KIF25 (NP_005346) produced in HEK293T cells. | |
0.1 mL | |
3834.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2F4 | |
Western Blot, Immunohistochemistry | |
kinesin family member 25 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction