Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF26B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100591
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100591 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100591 Supplier Invitrogen™ Supplier No. PA5100591
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KIF26B.

KIF26B is a member of kinesin-11 family and plays important role in embryogenesis. It is a downstream target of Sall1 and is essential for kidney development. KIF26B is also involved in tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF26B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Kif26b
Gene Accession No. Q2KJY2, Q7TNC6
Gene Alias 4832420M10; BC056349; D230039L06Rik; Kif26b; kinesin family member 26B; kinesin family protein 26b; kinesin-like protein KIF26B; N-11 kinesin; RGD1560022; RGD1560572
Gene Symbols Kif26b
Host Species Rabbit
Immunogen A synthesized peptide derived from human KIF26B(Accession Q2KJY2), corresponding to amino acid residues E1057-S1107.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 269152, 305012, 55083
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.