Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kif2a Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP255116
Description
Kif2a Polyclonal specifically detects Kif2a in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Kif2a | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
HK2, KIF2kinesin-2, kinesin heavy chain member 2, kinesin heavy chain member 2A, Kinesin, heavy chain, 2, Kinesin-2, kinesin-like protein KIF2A, KNS2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
KIF2A | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TAAGDVRPIMHHPPNQIDDLETQWGVGSSPQRDDLKLLCEQNEEEVSPQLFTFHEAVSQMVEMEEQVVEDHRAVFQESIRWLEDEKALLEMTEEVDYDVDSYATQLEAILEQKIDILTELRDKVKSFRAALQEEEQASKQINPKRPRA | |
100 μL | |
Cancer, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction | |
3796 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction