Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110684
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110684 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110684 Supplier Invitrogen™ Supplier No. PA5110684
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DNESVTVEWI ENGDTKGKEI DLESIFSLNP DLVPDEEIEP SPETPPPPAS SAKVNKIVKN RRTVASIKND PPSRDNRVVG SARARPSQFP EQSSSAQQNG SVSDISPVQA Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 92%.

KIF2A, a gene encoding the kinesin family member 2A protein, plays a pivotal role in neuronal development and function. This gene is part of the kinesin-13 subfamily, characterized by its unique ability to depolymerize microtubules, which are essential components of the cytoskeleton responsible for maintaining cell shape, enabling motility, and facilitating intracellular transport. KIF2A is particularly significant in neural settings, where it regulates microtubule dynamics crucial for neuronal migration and dendritic development. Dysfunctional KIF2A, due to mutations, has been linked to early-onset neurodegenerative diseases such as progressive microcephaly and mental disabilities due to impaired microtubule regulation, reflecting its foundational role in correct brain development and architecture. Additionally, KIF2A is involved in spindle assembly and chromosome segregation during cell division, further cementing its importance in both neurodevelopment and general cellular functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF2A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Kif2a
Gene Accession No. O00139, Q9WV63
Gene Alias C530030B14Rik; CDCBM3; HK2; hypothetical protein; KIF2; kif2a; kif2-a; kif2a {ECO:0000250; kif2a.L; kinesin family member 2A; kinesin heavy chain family, member 2; kinesin heavy chain member 2; kinesin heavy chain member 2A; kinesin heavy chain member 2A L homeolog; Kinesin, heavy chain, 2; kinesin-2; Kinesin-like protein KIF2A; kinesin-related protein XKIF2; Kns2; M-kinesin; RCJMB04_9a7; UniProtKB:O00139}; Unknown (protein for MGC:133806); XELAEV_18008243mg
Gene Symbols Kif2a
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human KIF2A. Recombinant protein control fragment (Product #RP-102374).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3796, 84391
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.