Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIF2A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540802
Description
Peptide sequence: IEPSPETPPP PASSAKVNKI VKNRRTVASI KNDPPSRDNR VVGSARARPS Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 79%; Rabbit: 93%; Rat: 93%.
Kinesins, such as KIF2, are microtubule-associated motor proteins. For background information on kinesins.
Specifications
KIF2A | |
Polyclonal | |
Unconjugated | |
Kif2a | |
C530030B14Rik; CDCBM3; HK2; hypothetical protein; KIF2; kif2a; kif2-a; kif2a {ECO:0000250; kif2a.L; kinesin family member 2A; kinesin heavy chain family, member 2; kinesin heavy chain member 2; kinesin heavy chain member 2A; kinesin heavy chain member 2A L homeolog; Kinesin, heavy chain, 2; kinesin-2; Kinesin-like protein KIF2A; kinesin-related protein XKIF2; Kns2; M-kinesin; RCJMB04_9a7; UniProtKB:O00139}; Unknown (protein for MGC:133806); XELAEV_18008243mg | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3796 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O00139 | |
Kif2a | |
Synthetic peptide directed towards the N-terminal of human KIF2A (aa 72-121). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction