Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIF2C Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11040688F
Description
KIF2C Polyclonal antibody specifically detects KIF2C in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
KIF2C | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
kinesin family member 2C, kinesin-like 6 (mitotic centromere-associated kinesin), Kinesin-like protein 6, kinesin-like protein KIF2C, MCAKKNSL6kinesin-like 6, Mitotic centromere-associated kinesin | |
The immunogen recognized by this antibody maps to a region between residue 50 and 100 of mouse Mitotic Centromere-Associated Kinesin using the numbering given in entry NP_608301.2 (GeneID 73804 ). | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, DNA Repair | |
11004 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FITC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction