Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF2C Antibody (KIF2C/4706), Novus Biologicals™
SDP

Catalog No. NB232994 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB232994 20 μg
NB234878 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB232994 Supplier Novus Biologicals Supplier No. NBP32055320UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KIF2C Monoclonal antibody specifically detects KIF2C in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen KIF2C
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KIF2C/4706
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL
Formulation 10mM PBS with 0.05% BSA
Gene Alias kinesin family member 2C, kinesin-like 6 (mitotic centromere-associated kinesin), Kinesin-like protein 6, kinesin-like protein KIF2C, MCAKKNSL6kinesin-like 6, Mitotic centromere-associated kinesin
Host Species Mouse
Immunogen Recombinant fragment (around aa500-700) of human KIF2C protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 11004
Target Species Human
Content And Storage Store at 4&drg;C. Do not freeze.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.