Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIF2C Antibody (KIF2C/6519), Novus Biologicals™

Mouse Monoclonal Antibody
$216.50 - $443.50
Specifications
Antigen | KIF2C |
---|---|
Clone | KIF2C/6519 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
Applications | Immunohistochemistry (Paraffin) |
Description
KIF2C Monoclonal antibody specifically detects KIF2C in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
KIF2C | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Cell Biology, Cellular Markers, DNA Repair | |
10mM PBS with 0.05% BSA | |
11004 | |
IgG2b κ | |
Protein A or G purified |
KIF2C/6519 | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human | |
kinesin family member 2C, kinesin-like 6 (mitotic centromere-associated kinesin), Kinesin-like protein 6, kinesin-like protein KIF2C, MCAKKNSL6kinesin-like 6, Mitotic centromere-associated kinesin | |
Recombinant fragment (around aa500-700) of human KIF2C protein (exact sequence is proprietary) | |
Primary | |
Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title