Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIF2C Antibody (OTI1C5), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271667MFV610
Description
KIF2C Monoclonal antibody specifically detects KIF2C in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Flow CytometrySpecifications
KIF2C | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Primate, Monkey | |
Purified |
Flow Cytometry | |
OTI1C5 | |
Flow Cytometry | |
kinesin family member 2C, kinesin-like 6 (mitotic centromere-associated kinesin), Kinesin-like protein 6, kinesin-like protein KIF2C, MCAKKNSL6kinesin-like 6, Mitotic centromere-associated kinesin | |
Full length human recombinant protein of human KIF2C(NP_006836) produced in HEK293T cell. | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, DNA Repair | |
11004 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction