Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIF2C Rabbit anti-Human, Clone: 7V9D2, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP316764100UL
This item is not returnable.
View return policy
Description
KIF2C Monoclonal antibody specifically detects KIF2C in Human samples. It is validated for Western BlotSpecifications
KIF2C | |
Monoclonal | |
Unconjugated | |
kinesin family member 2C, kinesin-like 6 (mitotic centromere-associated kinesin), Kinesin-like protein 6, kinesin-like protein KIF2C, MCAKKNSL6kinesin-like 6, Mitotic centromere-associated kinesin | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human KIF2C/KIF2C (Q99661). | |
100 μg | |
Cancer, Cell Biology, Cellular Markers, DNA Repair | |
11004 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
7V9D2 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction