Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIF3A Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP146837G
Description
KIF3A Polyclonal antibody specifically detects KIF3A in Human, Mouse samples. It is validated for Western BlotSpecifications
KIF3A | |
Polyclonal | |
Western Blot | |
KIF3, kinesin family member 3A, kinesin family protein 3A, kinesin-like protein KIF3A, Microtubule plus end-directed kinesin motor 3A | |
The immunogen for this product maps to a region between residue 652 and 702 of human kinesin family member 3A using the numbering given in entry AAC72294.1 (GeneID 11127). | |
0.1 mL | |
Cell Biology, Cytoskeleton Markers, Immunology, Neuroscience, Signal Transduction, Stem Cells | |
11127 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction