Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA120240
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA120240 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA120240 Supplier Invitrogen™ Supplier No. PA120240
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: PC12, rat cerebellum.

The kinesins constitute a large family of microtubule-dependent motor proteins which are responsible for the distribution of numerous organelles, vesicles and macromolecular complexes throughout the cell. Individual kinesin members play crucial roles in cell division, intracellular transport and membrane trafficking events including endocytosis and transcytosis. Members of the heterotrimeric kinesin II family of microtubule associated motors generally contain two different motor subunits from the KIF3 family, which includes KIF3A, B and C. KIF3 isoforms mediate anterograde transport of membrane bound organelles in neurons and melanosomes, transport between the endoplasmic reticulum and the Golgi, and transport of protein complexes within cilia and flagella required for their morphogenesis. KIF3A may influence neurogenesis at the level of embryonic cellular events, where the asymmetry of the genetic control circuit controlling left-right (L-R) axis determination is defined. Loss of KIF3A function in mice photoreceptors causes apoptotic cell death, suggesting that kinesin II mediated transport is required for proper cell fate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF3A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 5.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene KIF3A
Gene Accession No. P28741, Q9Y496
Gene Alias fc55h04; FLA10; hypothetical protein LOC550267; Kif3; KIF3A; Kifl; kinesin family member 3a; kinesin family protein 3A; kinesin-II subunit; kinesin-like protein KIF3A; Kinesin-like protein KIF3A-like protein; KLP-20; Kns3; microtubule plus end-directed kinesin motor 3A; N-4 kinesin; Unknown (protein for IMAGE:7984856); wu:fc55h04; zgc:110284; zgc:110284 protein
Gene Symbols KIF3A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminus of mouse KIF3A (amino acids 682-701) conjugated to KLH.
Purification Method IgG fraction
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11127, 16568, 84392
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.