Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIF3B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540775
Description
Peptide sequence: SAFSYICENE AIPMPPHWEN VNTQVPYQLI PLHNQTHEYN EVANLFGKTM Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 91%; Pig: 100%; Rabbit: 93%; Rat: 79%.
The PARP11 gene is part of the poly (ADP-ribose) polymerase family.
Specifications
KIF3B | |
Polyclonal | |
Unconjugated | |
KIF3B | |
AI854312; AW549267; FLA8; HH0048; KIAA0359; Kif3b; kinesin family member 3B; kinesin-like protein KIF3B; Kinesin-like protein KIF3B, N-terminally processed; KLP-11; microtubule plus end-directed kinesin motor 3B; mKIAA0359 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9371 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O15066 | |
KIF3B | |
Synthetic peptide directed towards the N-terminal of human PARP11. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction