Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kif3c Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | Kif3c |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Kif3c Polyclonal specifically detects Kif3c in Human samples. It is validated for Western Blot.Specifications
Kif3c | |
Polyclonal | |
Rabbit | |
O14782 | |
3797 | |
Synthetic peptides corresponding to KIF3C (kinesin family member 3C) The peptide sequence was selected from the middle region of KIF3C. Peptide sequence LQEQKERLEEEKAAIQDDRSLVSEEKQKLLEEKEKMLEDLRREQQATELL. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
KIF3C variant protein, kinesin family member 3C, kinesin-like protein KIF3C | |
KIF3C | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title