Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF3C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597899
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597899 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597899 Supplier Invitrogen™ Supplier No. PA597899
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KIF3C Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

KIF3C is a microtubule-based anterograde translocator for membranous organelles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF3C
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene KIF3C
Gene Accession No. O14782
Gene Alias KIF3C; KIF3C variant protein; kinesin family member 3C; kinesin-like protein KIF3C; mKIAA4058; N-4 kinesin
Gene Symbols KIF3C
Host Species Rabbit
Immunogen Recombinant Human Kinesin-like protein KIF3C protein (158-267AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3797
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.