Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kif4A Antibody [Alexa Fluor« 647], Novus Biologicals Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP335106AF647
Description
Kif4A Polyclonal antibody specifically detects Kif4A in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ ImmunofluorescenceSpecifications
Kif4A | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
Alexa Fluor 647 | |
chromokinesin-A, chromosome-associated kinesin KIF4A, FLJ12530, FLJ12655, FLJ14204, FLJ20631, HSA271784, KIF4G1, KIF4KIF4-G1, kinesin family member 4A | |
Recombinant fusion protein containing a sequence corresponding to amino acids 870-1080 of human Kif4A (NP_036442.3).,, Sequence:, LIGELVSSKIQVSKLESSLKQSKTSCADMQKMLFEERNHFAEIETELQAELVRMEQQHQEKVLYLLSQLQQSQMAEKQLEESVSEKEQQLLSTLKCQDEELEKMREVCEQNQQLLRENEIIKQKLTLLQVASRQKHLPKDTLLSPDSSFEYVPPKPKPSRVKEKFLEQSMDIEDLKYCSEHSVNEHEDGDGDDDEGDDEEWKPTKLVKVSR | |
0.1 mL | |
Core ESC Like Genes, Lipid and Metabolism, Mitotic Regulators, Neuroscience, Stem Cell Markers | |
24137 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction