Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530492
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530492 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530492 Supplier Invitrogen™ Supplier No. PA530492
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, HepG2, HCT116, 293T, A431. Predicted reactivity: Xenopus laevis (80%), Chicken (81%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the kinesin 4 or subfamily of kinesin related proteins. This subfamily is also referred to as the chromokinesins. The encoded protein is an ATP dependent microtubule-based motor protein that is involved in the intracellular transport of membranous organelles. This protein also associates with condensed chromosome arms and may be involved maintaining chromosome integrity during mitosis. This protein may also be involved in the organization of the central spindle prior to cytokinesis. A pseudogene of this gene is found on chromosome X.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF4A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Kif4a
Gene Accession No. O95239
Gene Alias AI323435; Chromokinesin; chromokinesin-A; Chromosome-associated kinesin KIF4; chromosome-associated kinesin KIF4A; D330050K22Rik; FLJ12530; FLJ12655; FLJ14204; FLJ20631; HSA271784; KIF4; KIF4A; KIF4G1; KIF4-G1; kinesin family member 4; kinesin family member 4A; kinesin heavy chain member 4; Kns4; MRX100; N-5 kinesin
Gene Symbols Kif4a
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human KIF4A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24137
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.