Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587703
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587703 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587703 Supplier Invitrogen™ Supplier No. PA587703
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LIGELVSSKI QVSKLESSLK QSKTSCADMQ KMLFEERNHF AEIETELQAE LVRMEQQHQE KVLYLLSQLQ QSQMAEKQLE ESVSEKEQQL LSTLKCQDEE LEKMREVCEQ NQQLLRENEI IKQKLTLLQV ASRQKHLPKD TLLSPDSSFE YVPPKPKPSR VKEKFLEQSM DIEDLKYCSE HSVNEHEDGD GDDDEGDDEE WKPTKLVKVS R; Positive Samples: U-87MG, LO2, 293T, SW480; Cellular Location: Chromosome, Cytoplasm, Midbody, Nucleus matrix, cytoskeleton, spindle.

This gene encodes a member of the kinesin 4 or subfamily of kinesin related proteins. This subfamily is also referred to as the chromokinesins. The encoded protein is an ATP dependent microtubule-based motor protein that is involved in the intracellular transport of membranous organelles. This protein also associates with condensed chromosome arms and may be involved maintaining chromosome integrity during mitosis. This protein may also be involved in the organization of the central spindle prior to cytokinesis. A pseudogene of this gene is found on chromosome X.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF4A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Kif4a
Gene Accession No. O95239, P33174
Gene Alias AI323435; Chromokinesin; chromokinesin-A; Chromosome-associated kinesin KIF4; chromosome-associated kinesin KIF4A; D330050K22Rik; FLJ12530; FLJ12655; FLJ14204; FLJ20631; HSA271784; KIF4; KIF4A; KIF4G1; KIF4-G1; kinesin family member 4; kinesin family member 4A; kinesin heavy chain member 4; Kns4; MRX100; N-5 kinesin
Gene Symbols Kif4, Kif4a
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 870-1080 of human KIF4A (NP_0364423).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16571, 24137, 84393
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.