Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF5B Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501567386
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-7386 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7386 Supplier Bethyl Laboratories, Inc Supplier No. A304306A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat

Kinesin belongs to the group of microtubule-associatied motor proteins known to convert chemical energy released from nucleoside triphosphates (preferentially from ATP) into mechanical energy. Conventional kinesin, member of kinesin superfamily comprising more than 100 proteins, is involved in the anterograde vesicle transport in neuronal cells. Kinesin purified from mammalian brain homogenates is a heterotetramer consisting of two heavy (120 to 130 kDa) and two light chains (60 to 70 kDa), resulting in a molecular mass about 400 kDa. Each heavy chain contains an N-terminal globular motordomain with both a microtuble-binding site and an ATPase active center, stalk region responsible for heavy chain dimerization and finally C-terminal globular tail domain, which is implicated in cargo binding. Light chains may have a regulatory function.

Specifications

Antigen KIF5B
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene KIF5B
Gene Accession No. P33176, Q61768
Gene Alias KINH, KNS, KNS1, UKHC
Gene Symbols Kif5b
Host Species Rabbit
Immunogen Between 675 and 725
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16573, 3799
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.