Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIF6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | KIF6 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
KIF6 Polyclonal specifically detects KIF6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
KIF6 | |
Polyclonal | |
Rabbit | |
Human | |
C6orf102, chromosome 6 open reading frame 102, dJ1043E3.1, dJ137F1.4, dJ188D3.1, DKFZp451I2418, kinesin family member 6, kinesin-like protein KIF6, MGC33317 | |
KIF6 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
221458 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:NEAVLNEEINPRLVIKRLQKEIQELKDELAMVTGEQRTEALTEAELLQLEKLITSFLEDQDSDSRLEVGADMRKVHHCFHHL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title