Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF6 Monoclonal Antibody (OTI5G2)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527539

Catalog No. PIMA527539


Only null left
Add to Cart

Description

Description

KIF6 Monoclonal Antibody for Western Blot, IHC (P)

KIF6 belongs to the kinesin-like protein family. There are four named isoforms. The KIF6 719Arg variant has been associated with an increased risk of coronary disease and an increased response to statin therapy.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

KIF6
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q6ZMV9
KIF6
Human recombinant protein fragment corresponding to amino acids 1-350 of KIF6 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
OTI5G2
Unconjugated
KIF6
C6orf102; D130004B10Rik; D130084M03Rik; dJ1043E3.1; dJ137F1.4; dJ188D3.1; KIF6; kinesin 6; kinesin family member 6; kinesin-like protein KIF6; kinesin-related protein 3; Krp3
Mouse
Affinity chromatography
RUO
221458
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.