Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566190
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566190 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566190 Supplier Invitrogen™ Supplier No. PA566190
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RKHQQGIYSID EDEKLIPSLE IILPRDLADG FVNNKRESYK FKFQRIFDQD ANQETVFENI AKPVAGSVL Highest antigen sequence identity to the following orthologs - mouse 84%, rat 84%.

KIF6 belongs to the kinesin-like protein family. There are four named isoforms. The KIF6 719Arg variant has been associated with an increased risk of coronary disease and an increased response to statin therapy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF6
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIF6
Gene Accession No. Q6ZMV9
Gene Alias C6orf102; D130004B10Rik; D130084M03Rik; dJ1043E3.1; dJ137F1.4; dJ188D3.1; KIF6; kinesin 6; kinesin family member 6; kinesin-like protein KIF6; kinesin-related protein 3; Krp3
Gene Symbols KIF6
Host Species Rabbit
Immunogen Recombinant Human KIF6. Recombinant protein control fragment (Product #RP-106875).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221458
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.