Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560258
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560258 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560258 Supplier Invitrogen™ Supplier No. PA560258
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MWINQELKQK LGGVNAVGHS RGGEKRSLCS EGRQAPGNED ELHLAPELLW LSPLTEGAPR TREETRDLVH APLPLTWKRS SLCGEEQGSP EELRQREAAE PLVGRVLPVG E Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 69%.

Vasoactive intestinal peptide is a neuroprotective factor that has a stimulatory effect on the growth of some tumor cells and an inhibitory effect on others. This gene encodes a protein that is upregulated by vasoactive intestinal peptide and may be involved in its stimulatory effect on certain tumor cells. The encoded protein contains one homeobox and nine zinc finger domains, suggesting that it functions as a transcription factor. This gene is also upregulated in normal proliferative tissues. Finally, the encoded protein may increase the viability of certain cell types through modulation of p53 activity. Alternatively spliced transcript variants encoding the same protein have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIF7
Gene Accession No. Q2M1P5
Gene Alias ACLS; AGBK; EQYK340; HLS2; JBTS12; KIF7; kinesin 7; kinesin family member 7; kinesin-like protein KIF7; UNQ340; UNQ340/PRO539
Gene Symbols KIF7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIF7. Recombinant protein control fragment (Product #RP-98154).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374654
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.