Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIF7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578678
Description
Positive Control: 293T, A431, HeLa, HepG2 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Vasoactive intestinal peptide is a neuroprotective factor that has a stimulatory effect on the growth of some tumor cells and an inhibitory effect on others. This gene encodes a protein that is upregulated by vasoactive intestinal peptide and may be involved in its stimulatory effect on certain tumor cells. The encoded protein contains one homeobox and nine zinc finger domains, suggesting that it functions as a transcription factor. This gene is also upregulated in normal proliferative tissues. Finally, the encoded protein may increase the viability of certain cell types through modulation of p53 activity. Alternatively spliced transcript variants encoding the same protein have been described.
Specifications
| KIF7 | |
| Polyclonal | |
| Unconjugated | |
| KIF7 | |
| ACLS; AGBK; EQYK340; HLS2; JBTS12; KIF7; kinesin 7; kinesin family member 7; kinesin-like protein KIF7; UNQ340; UNQ340/PRO539 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 374654 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 2.03 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q2M1P5 | |
| KIF7 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human KIF7. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction