Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIFC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554359
Description
Immunogen sequence: NIRVIARVRP VTKEDGEGPE ATNAVTFDAD DDSIIHLLHK GKPVSFELDK VFSPQASQQD VFQEVQALVT SCIDGFNVCI FAYGQT Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
KIFC3 belongs to the large superfamily of kinesins, molecular motors that use the energy of ATP hydrolysis to translocate cargoes along microtubules. Members share extensive homology within a globular domain containing the microtubule- and ATP-binding sites and have a coiled-coil stalk domain that mediates oligomerization. Different kinesin family members participate in specific and diverse motile processes, such as cell division, organelle transport, and nuclear movement (Hoang et al., 1998 ).
Specifications
| KIFC3 | |
| Polyclonal | |
| Unconjugated | |
| KIFC3 | |
| AI325457; BB123200; DKFZp686D23201; KIFC3; kinesin family member C3; kinesin-like protein KIFC3; KRP4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3801 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BVG8 | |
| KIFC3 | |
| Recombinant protein corresponding to Human KIFC3. Recombinant protein control fragment (Product #RP-91744). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction