Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Kifunensine
GSA_VAGREENER_CHOICESDP

Catalog No. 32071 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-071 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 32-071 Supplier R&D Systems™ Supplier No. 3207/1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibitor of class I α-mannosidases

Kifunensine is an inhibitor of class I α-mannosidases that inhibits glycoprotein processing. Kifunensine inhibits human endoplasmic reticulum α-1,2-mannosidase I and Golgi Class I mannosidases IA, IB and IC with Ki values of 130 and 23 nM respectively. Kifunensine disrupts normal trafficking of the GLUT1 transporter, leading to decreased glucose uptake in human renal epithelial cells. Kifunensine impaires tumour cell aggregation in human ovarian cancer cells in vitro.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 109944-15-2
Molecular Formula C8H12N2O6

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 232.19
Purity >98%
Molecular Weight (g/mol) 232.192
Chemical Name or Material Kifunensine
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.