Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ KIMAX™-35 300mL BOD Bottles
Shop All DWK Life Sciences Products

Click to view available options
No. Sequence:
1 to 24
Unnumbered
Stopper:
With stopper
Without stopper
Description
For sampling and incubation of aqueous samples for biochemical oxygen demand (BOD) analysis
Flared mouth forms a water seal to prevent air from being drawn into the bottle during incubation, while shoulder radius provides an interior shape that sweeps entrapped air out of the stopper opening.
- Type 1, Class B borosilicate glass
- Available numbered and unnumbered With glass stopper unless otherwise noted
- Optional snap lid secures stopper in bottle and reduces evaporation of water seal during incubation (sold separately, 02-939)
- Size: 2.75 dia. × 6.5 in. H (69 × 165mm); neck I.D. approximately 0.66 in. (16.7mm)
Order Info
Plastic snap lids not included. For these, see Cat. No. 02-939.
Compliance
Conforms to ASTM™ E 438
Specifications
Specifications
| Certifications/Compliance | ASTM E438 type I, class A |
| Volume (Metric) | 300 mL |
| For Use With (Application) | For sampling and incubation of aqueous samples |
| Material | Borosilicate Glass |
| No. Sequence | Unnumbered |
| Stopper | Without stopper |
| Volume (English) | 10.14 oz. |
| Height (English) | 6.5 in. |
| Height (Metric) | 16.5 cm |
| Diameter (English) | 2.7 in. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction