Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ KIMAX™ Reagent Bottles
SDP

Catalog No. 028868G Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
10,000 mL
1000 mL
20,000 mL
2000 mL
Diameter (English) Inner Neck:
1.06 in.
2.28 in.

For accurate rubber stopper fit

  • Borosilicate glass
  • Tooled top and narrow mouth

Order Info

Stoppers are available separately; order online

Compliance

Meet ASTM™ E 438-92, Type I, Class A requirements.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 338.1 oz., 10000mL
Capacity (Metric) 10,000 mL
Capacity (English) 338.1 oz.
Diameter (English) 9.05 in.
Diameter (English) Inner Neck 2.28 in.
Diameter (Metric) Inner Neck 58 mm
Includes Tooled neck
Material Borosilicate Glass
Stopper Number 12
Certifications/Compliance ASTM E438-92, Type I, Class A
Mouth Narrow
Height (Metric) 400 mm
Height (English) 15.74 in.
Closure Material Rubber
Diameter (Metric) 230 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.