Learn More
Ansell FluidShield™ Procedure Mask
Catalog No. 189994800 Shop All Ansell Products
Change view
Click to view available options
X-Large
Catalog No.
18-999-4800 X-Large
1 options
This item is not returnable. View return policy
Catalog No. 18-999-4800 Supplier Ansell Supplier No. 47107
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Procedure mask is designed to provide the maximum level of fluid resistance recognized by ASTM F2100-11.
  • For heavy fluid exposure
  • Pleated
  • Ear loops tie for easy donning
  • Made with a breathable film and So Soft lining
  • Constructed with a combination of materials to resist fluid penetration upon impact
  • Fog-free
  • Foam strip for extra comfort
  • ASTM F2100-11 Level 3 rating for heavy fluid protection
TRUSTED_SUSTAINABILITY
Specifications
Product Type Medical/Procedural Face Mask
Color Orange
Fastening Type Earloops
Style Pleated
Filter Efficiency 0.99
Model 47107
Pore Size 0.1 μm (PFE), 3 μm (BFE)