Learn More
Ansell KleenGuard™ G60 Polyurethane Coated Cut-Resistant Gloves, ANSI Level A2
Shop All Ansell ProductsDescription
- Offer EN388 Level 3 / ANSI Level 2 Cut Resistance and CPPT = 686 grams
- Made of ultra-high molecular weight polyethylene (UHMWPE) yarn with nylon and Lycra fiber, which provides both cut resistance and a comfortable fit
- Polyurethane coating enhances grip and the dark color helps hide dirt for longer wear
- Each pair has a dedicated right and left glove
Specifications
Specifications
| Coating Material | Polyurethane |
| Color | Black, White |
| Cut Resistance Rating | ANSI A2 |
| Material | Dyneema, Fiber, Lycra |
| Product Type | Cut-resistant Gloves |
| Size | Medium |
| EN388 Standard | EN 388 Cut Level 3 |
| For Use With (Application) | Metal fabrication, Glass handling, aerospace and automotive assembly |
| Glove Cuff Style | Knit |
| Model | 38690 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.