Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ G60 Polyurethane Coated Cut Resistant Gloves, ANSI Level A4
GREENER_CHOICE

Catalog No. 17986023 Shop All Ansell Products
Change view
Click to view available options
Glove Cuff Color:
Black
Brown
Gray
Navy
Purple
Size:
2X-Large, 11
Large, 9
Medium, 8
Small, 7
X-Large, 10
Length (English):
9.4 in.
9.8 in.
10.2 in.
10.6 in.
11 in.
Length (Metric):
24 cm
25 cm
26 cm
27 cm
28 cm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Glove Cuff Color Size Length (English) Length (Metric)
17-986-023 Black X-Large, 10 10.6 in. 27 cm
17-986-020 Navy Small, 7 9.4 in. 24 cm
17-986-021 Gray Medium, 8 9.8 in. 25 cm
17-986-024 Brown 2X-Large, 11 11 in. 28 cm
17-986-022 Purple Large, 9 10.2 in. 26 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-986-023 Supplier Ansell Supplier No. 98238
Add to Cart
Add to Cart
This item is not returnable. View return policy

Knit gloves with polyurethane coating feature dexterity and a secure grip in dry and light oily conditions, and are ideal when encountering medium cut hazards.

  • EN Level 5 cut resistance Spun Dyneema™ fiber with glass, stainless steel and spandex
  • Black polyurethane coating enhances grip
  • Dark coating hides dirt for longer wear
  • ANSI Level 4/EN388
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Construction Seamless Knit
Coating Material Polyurethane
Color Black, Salt and Pepper
Cut Resistance Rating ANSI A4
Material Dyneema, Polyurethane, Spandex, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size X-Large, 10
EN388 Standard EN 388 Cut Level 5
For Use With (Application) Metal fabrication, Glass handling, handling sharp parts, automotive assembly
Glove Cuff Color Black
Model 98238
Puncture Rating ANSI/ISEA 105-16 Level 3
Series G60
Yarn Gauge 13
Length (English) 10.6 in.
Length (Metric) 27 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.