Learn More
Ansell KleenGuard™ V30 Nemesis™ Small Safety Eyewear
Get everything you love about the original KleenGuard Nemesis Safety Eyewear - stylish good looks and durable, lightweight construction – but in with smaller dimensions.
Supplier: Ansell 38480
View more versions of this product
Description
- Durable, lightweight construction
- Offer a sport design that fits smaller, slimmer heads without adjustments
- Features a stronger nose piece and a wider bridge for enhanced comfort
- Polycarbonate lenses that provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss
- Wraparound frame has a base curve of 8 that extends the range of peripheral protection
- Each pair comes with a neck cord for securing, so they are ready at a moment’s notice
- Manufactured to comply with the ANSI / ISEA Z87.1+ standard for personal eye protection
- Feature a sporty, flexible, lightweight design
- Patented FLEX-DRY designed to be extremely flexible, to channel sweat away from the eyes, and to help prevent slippage down the nose
Specifications
Hardcoat | |
Meets ANSI Z87.1+ standards for high impact (38480-00 and 38480-01 meets ANSI Z87.1+2010 and 38480-02 meets ANSI Z87.1+2015) | |
Safety Eyewear | |
Black frame with green tips |
Indoor/outdoor | |
Indoor/Outdoor | |
Black |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.