Learn More
Ansell KleenGuard™ V30 Nemesis™ Small Safety Eyewear
Get everything you love about the original KleenGuard Nemesis Safety Eyewear - stylish good looks and durable, lightweight construction – but in with smaller dimensions.
$15.51 - $19.52
Catalog Number | Mfr. No. | Frame Color | Frame Type | Lens Coating | Lens Tint | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Frame Color | Frame Type | Lens Coating | Lens Tint | Price | Quantity & Availability | ||
19-169-470
![]() |
Ansell
38474 |
Black | Black frame with black tips | Anti-fog | Amber |
Each for $15.51
Case of 12 EA for $186.17
|
|
||
19-169-472
![]() |
Ansell
38478 |
Black | Black frame with pink tips | Anti-fog | Clear |
Each for $19.52
Dozen of 12 EA for $234.27
|
![]() |
||
19-169-471
|
Ansell
38476 |
Black | Black frame with black tips | Hardcoat | Smoke | N/A | N/A | N/A | |
19-169-473
|
Ansell
38480 |
Black | Black frame with green tips | Hardcoat | Indoor/Outdoor | N/A | N/A | N/A | |
Description
- Durable, lightweight construction
- Offer a sport design that fits smaller, slimmer heads without adjustments
- Features a stronger nose piece and a wider bridge for enhanced comfort
- Polycarbonate lenses that provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss
- Wraparound frame has a base curve of 8 that extends the range of peripheral protection
- Each pair comes with a neck cord for securing, so they are ready at a moment’s notice
- Manufactured to comply with the ANSI / ISEA Z87.1+ standard for personal eye protection
- Feature a sporty, flexible, lightweight design
- Patented FLEX-DRY designed to be extremely flexible, to channel sweat away from the eyes, and to help prevent slippage down the nose
Specifications
Safety Eyewear | |
Black |